Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01408800_m1
          See other TFCP2L1 GE Assays ›
          Gene Symbol:
          Tfcp2l1
          Gene Name:
          transcription factor CP2-like 1
          Gene Aliases:
          Tcfcp2l1
          Chromosome Location:
          Chr.13: 34610689 - 34671617 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01408800_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfcp2l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107170.1 NP_001100640.1 14 - 15 1476 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000121.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          epithelial cell maturation
          regulation of transcription from RNA polymerase II promoter
          cytoplasm organization
          salivary gland development
          determination of adult lifespan
          positive regulation of growth
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P140079 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P977318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114504 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73686 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P140079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P977318 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P977318 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P140079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P140079 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P977318 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P140079 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8698 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763535 - -
          Affymetrix Rat Gene 1.0 ST Array 10763538 - -
          Affymetrix Rat Gene 1.0 ST Array 10763546 - -
          Affymetrix Rat Gene 1.0 ST Array 10763545 - -
          Affymetrix Rat Gene 1.0 ST Array 10763532 - -
          Affymetrix Rat Gene 1.0 ST Array 10763533 - -
          Affymetrix Rat Gene 1.0 ST Array 10763542 - -
          Affymetrix Rat Gene 1.0 ST Array 10763544 - -
          Affymetrix Rat Gene 1.0 ST Array 10763540 - -
          Affymetrix Rat Gene 1.0 ST Array 10763536 - -
          Affymetrix Rat Gene 1.0 ST Array 10763541 - -
          Affymetrix Rat Gene 1.0 ST Array 10763539 - -
          Affymetrix Rat Gene 1.0 ST Array 10763534 - -
          Affymetrix Rat Gene 1.0 ST Array 10763537 - -
          Affymetrix Rat Gene 1.0 ST Array 10763543 - -
          Affymetrix Rat Gene 1.1 ST Array 10763531 - -
          Rat Genome 230 2.0 Array 1393337_PM_at - -
          Rat Genome 230 2.0 Array 1393337_at - -
          Affymetrix Rat Clariom S Assay PSR1300000669.rn.1 TC1300000121.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000674.rn.1 TC1300000121.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000668.rn.1 TC1300000121.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000676.rn.1 TC1300000121.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000673.rn.1 TC1300000121.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300000675.rn.1 TC1300000121.rn.1 RTA TC
          Rat Expression Set 230 1393337_PM_at - -
          Rat Expression Set 230 1393337_at - -
          Affymetrix Rat Gene 1.0 ST Array 10763531 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21114186 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363428 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline