Search
Search
| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013197.1 | NP_037329.1 | 5 - 6 | 838 | 79 | |
| GenBank mRNA | BC081857.1 | - | 859 | 7133382 | 79 | |
| D86297.1 | - | 5 - 6 | 838 | 79 | ||
Set Membership: |
|