Search
Search
| Gene Symbol: Crhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022714.1 | NP_073205.1 | 10 - 11 | 1267 | 103 | |
| XM_006236556.2 | XP_006236618.1 | 11 - 12 | 1257 | 103 | ||
| XM_006236557.2 | XP_006236619.1 | 11 - 12 | 1254 | 103 | ||
| XM_006236558.1 | XP_006236620.1 | 10 - 11 | 1285 | 103 | ||
| GenBank mRNA | U16253.1 | - | 10 - 11 | 1267 | 103 | |
Set Membership: |
|