Search
Search

Hnrnpa2b1| Gene Symbol: Hnrnpa2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104613.1 | NP_001098083.1 | 5 - 6 | 617 | 103 | |
| GenBank mRNA | AB006815.1 | - | 4 - 5 | 581 | 103 | |
| AB006816.1 | - | 5 - 6 | 617 | 103 | ||
| AB006817.1 | - | 4 - 5 | 581 | 103 | ||
| AB006818.1 | - | 5 - 6 | 617 | 103 | ||
| BC166548.1 | - | 212 | 103 | |||
Set Membership: |
|