Search
Search
| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080890.1 | NP_543166.1 | 2 - 3 | 74 | 73 | |
| GenBank mRNA | AF393243.1 | - | 2 - 3 | 74 | 73 | |
| BC061533.1 | - | 2 - 3 | 98 | 6918024 | 73 | |
Set Membership: |
|