Search
Search
| Gene Symbol: Erlin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106353.1 | NP_001099823.1 | 6 - 7 | 527 | 118 | |
| XM_006231441.2 | XP_006231503.1 | 6 - 7 | 547 | 118 | ||
| GenBank mRNA | BC161938.1 | - | 6 - 7 | 479 | 9022977 | 118 |
| FQ209728.1 | - | 6 - 7 | 548 | 118 | ||
| FQ221472.1 | - | 5 - 6 | 412 | 118 | ||
Set Membership: |
|