Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01411506_m1
          See other TGFB2 GE Assays ›
          Gene Symbol:
          Tgfb2
          Gene Name:
          transforming growth factor, beta 2
          Gene Aliases:
          TGF-B2
          Chromosome Location:
          Chr.13: 105039639 - 105141689 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01411506_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031131.1 NP_112393.1 7 - 8 1473 144
          XM_006250448.2 XP_006250510.1 6 - 7 2299 144
          GenBank mRNA AF135598.1 - 6 - 7 1083 144
          AF153012.2 - 7 - 8 1473 144
          AF153013.1 - 6 - 7 1389 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002154.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgfb2

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          skeletal system development
          cartilage condensation
          regulation of cell growth
          blood vessel development
          eye development
          response to hypoxia
          kidney development
          epithelial to mesenchymal transition
          hair follicle development
          blood vessel remodeling
          heart morphogenesis
          heart valve morphogenesis
          protein phosphorylation
          cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          SMAD protein import into nucleus
          axon guidance
          salivary gland morphogenesis
          heart development
          skeletal muscle tissue development
          female pregnancy
          aging
          cell death
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          glial cell migration
          response to radiation
          response to cold
          response to wounding
          cardioblast differentiation
          response to organonitrogen compound
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          negative regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of macrophage cytokine production
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell growth
          cell migration
          signal transduction by protein phosphorylation
          hemopoiesis
          extracellular matrix organization
          collagen fibril organization
          positive regulation of cell growth
          negative regulation of cell growth
          lung development
          positive regulation of cell migration
          neutrophil chemotaxis
          thyroid gland development
          hindbrain development
          pancreas development
          hair follicle morphogenesis
          activation of protein kinase activity
          response to estradiol
          response to retinoic acid
          response to progesterone
          positive regulation of stress-activated MAPK cascade
          regulation of transforming growth factor beta2 production
          response to vitamin D
          positive regulation of cell adhesion mediated by integrin
          response to cytokine
          response to laminar fluid shear stress
          wound healing
          regulation of cell proliferation
          dopamine biosynthetic process
          response to drug
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of neuron apoptotic process
          response to estrogen
          cell-cell junction organization
          positive regulation of integrin biosynthetic process
          positive regulation of ossification
          positive regulation of cell cycle
          positive regulation of heart contraction
          somatic stem cell division
          cell development
          digestive tract development
          embryonic digestive tract development
          neuron fate commitment
          neuron development
          embryonic neurocranium morphogenesis
          inner ear development
          negative regulation of epithelial cell proliferation
          positive regulation of protein secretion
          negative regulation of immune response
          positive regulation of immune response
          negative regulation of release of sequestered calcium ion into cytosol
          positive regulation of cell division
          positive regulation of catagen
          positive regulation of cardioblast differentiation
          cardiac muscle cell proliferation
          cardiac epithelial to mesenchymal transition
          face morphogenesis
          frontal suture morphogenesis
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          negative regulation of cartilage development
          positive regulation of activation-induced cell death of T cells
          extrinsic apoptotic signaling pathway
          regulation of extracellular matrix organization
          regulation of complement-dependent cytotoxicity
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          beta-amyloid binding
          receptor signaling protein serine/threonine kinase activity
          receptor binding
          type II transforming growth factor beta receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          growth factor activity
          type III transforming growth factor beta receptor binding
          protein homodimerization activity
          protein heterodimerization activity
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P236342 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12532 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163532 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11728 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50012 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109531 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12532 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P236342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109531 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12532 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P236342 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P236342 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109531 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12532 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P246538 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701290975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97420 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83323 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54315 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37929 - -
          Agilent Rat cDNA Microarray (G4105A) 701290975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770587 - -
          Affymetrix Rat Gene 1.0 ST Array 10770585 - -
          Affymetrix Rat Gene 1.0 ST Array 10770580 - -
          Affymetrix Rat Gene 1.0 ST Array 10770581 - -
          Affymetrix Rat Gene 1.0 ST Array 10770586 - -
          Affymetrix Rat Gene 1.0 ST Array 10770579 - -
          Affymetrix Rat Gene 1.0 ST Array 10770578 - -
          Affymetrix Rat Gene 1.0 ST Array 10770584 - -
          Affymetrix Rat Gene 1.0 ST Array 10770583 - -
          Affymetrix Rat Gene 1.0 ST Array 10770582 - -
          Affymetrix Rat Gene 1.1 ST Array 10770577 - -
          Rat Genome 230 2.0 Array 1376425_PM_at - -
          Rat Genome 230 2.0 Array 1388011_PM_a_at - -
          Rat Genome 230 2.0 Array 1392382_PM_at - -
          Rat Genome 230 2.0 Array 1387172_a_at - -
          Rat Genome 230 2.0 Array 1388011_a_at - -
          Rat Genome 230 2.0 Array 1387172_PM_a_at - -
          Rat Genome 230 2.0 Array 1378963_PM_at - -
          Rat Genome 230 2.0 Array 1378963_at - -
          Rat Genome 230 2.0 Array 1392382_at - -
          Rat Genome 230 2.0 Array 1376425_at - -
          Affymetrix HT Rat Focus Array 1388011_a_at - -
          Affymetrix HT Rat Focus Array 1388011_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387172_a_at - -
          Affymetrix HT Rat Focus Array 1376425_at - -
          Affymetrix HT Rat Focus Array 1378963_PM_at - -
          Affymetrix HT Rat Focus Array 1392382_at - -
          Affymetrix HT Rat Focus Array 1392382_PM_at - -
          Affymetrix HT Rat Focus Array 1387172_PM_a_at - -
          Affymetrix HT Rat Focus Array 1376425_PM_at - -
          Affymetrix HT Rat Focus Array 1378963_at - -
          Affymetrix Rat Clariom S Assay PSR1300015382.rn.1 TC1300002154.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300015383.rn.1 TC1300002154.rn.1 RTA TC
          Rat Expression Set 230 1388011_a_at - -
          Rat Expression Set 230 1376425_PM_at - -
          Rat Expression Set 230 1388011_PM_a_at - -
          Rat Expression Set 230 1376425_at - -
          Rat Expression Set 230 1387172_a_at - -
          Rat Expression Set 230 1387172_PM_a_at - -
          Rat Expression Set 230 1392382_at - -
          Rat Expression Set 230 1378963_PM_at - -
          Rat Expression Set 230 1378963_at - -
          Rat Expression Set 230 1392382_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770577 - -
          Rat Genome U34 Set M96643_at - -
          Rat Genome U34 Set rc_AA963004_at - -
          Rat Genome U34 Set rc_AI231342_at - -
          Rat Genome U34 Set M96643_at - -
          Rat Genome U34 Set M96643_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22350958 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373682 - -
          RatRef-12 Expression BeadChip ILMN_69134 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline