Search
Search

Pex19
LOC100911356| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107375.2 | NP_001100845.1 | 6 - 7 | 801 | 87 | |
| GenBank mRNA | CO573662.1 | - | 1 - 2 | 105 | 87 | |
| Y09049.1 | - | 6 - 7 | 780 | 87 | ||
| Gene Symbol: LOC100911356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751804.3 | XP_003751852.1 | 6 - 7 | 794 | 87 | |
Set Membership: |
|