Search
Search
| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013079.1 | NP_001013097.1 | 11 - 12 | 1284 | 74 | |
| XM_006235816.2 | XP_006235878.1 | 12 - 13 | 1533 | 74 | ||
| GenBank mRNA | BC091208.1 | - | 11 - 12 | 1284 | 7381353 | 74 |
Set Membership: |
|