Search
Search
| Gene Symbol: Slc4a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053424.1 | NP_445876.1 | 9 - 10 | 1101 | 75 | |
| XM_006250788.2 | XP_006250850.1 | 9 - 10 | 1561 | 75 | ||
| XM_006250789.2 | XP_006250851.1 | 9 - 10 | 1561 | 75 | ||
| XM_006250790.2 | XP_006250852.1 | 10 - 11 | 1279 | 75 | ||
| XM_006250791.2 | XP_006250853.1 | 9 - 10 | 1113 | 75 | ||
| GenBank mRNA | AF004017.1 | - | 6 - 7 | 939 | 75 | |
| AF027362.1 | - | 6 - 7 | 971 | 75 | ||
| AF124441.1 | - | 9 - 10 | 1101 | 75 | ||
| AF210250.1 | - | 9 - 10 | 1108 | 75 | ||
| AF254802.1 | - | 9 - 10 | 1101 | 75 | ||
Set Membership: |
|