Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414317_m1
          See other SALL2 GE Assays ›
          Gene Symbol:
          Sall2
          Gene Name:
          spalt-like transcription factor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 28728471 - 28746042 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414317_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sall2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107262.1 NP_001100732.1 1 - 2 667 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001588.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          eye development
          biological_process
          neural tube development
          negative regulation of transcription, DNA-templated
          molecular_function
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105152 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105157 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105157 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105157 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105152 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P185027 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783347 - -
          Affymetrix Rat Gene 1.0 ST Array 10783348 - -
          Affymetrix Rat Gene 1.0 ST Array 10783345 - -
          Affymetrix Rat Gene 1.0 ST Array 10783344 - -
          Affymetrix Rat Gene 1.1 ST Array 10783343 - -
          Rat Genome 230 2.0 Array 1389437_at - -
          Rat Genome 230 2.0 Array 1389437_PM_at - -
          Affymetrix HT Rat Focus Array 1389437_PM_at - -
          Affymetrix HT Rat Focus Array 1389437_at - -
          Affymetrix Rat Clariom S Assay PSR1500010242.rn.1 TC1500001588.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500010244.rn.1 TC1500001588.rn.1 RTA TC
          Rat Expression Set 230 1389437_PM_at - -
          Rat Expression Set 230 1389437_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783343 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21362086 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362246 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline