Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01414745_m1
          See other RNASE4 GE Assays ›
          Gene Symbol:
          Rnase4
          Gene Name:
          ribonuclease, RNase A family 4
          Gene Aliases:
          Rab1
          Chromosome Location:
          Chr.15: 28018040 - 28035395 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01414745_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnase4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020082.2 NP_064467.1 1 - 2 46 64
          GenBank mRNA BC070953.1 - 1 - 2 46 7103267 64

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC070953.1, NM_020082.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnase4

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to starvation
          nucleic acid phosphodiester bond hydrolysis
          nucleic acid binding
          endonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180507 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12059 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12059 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P369206 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12059 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700483154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84669 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11139 - -
          Agilent Rat cDNA Microarray (G4105A) 700483154 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10779833 - -
          Affymetrix Rat Gene 1.0 ST Array 10779834 - -
          Affymetrix Rat Gene 1.1 ST Array 10779832 - -
          Rat Genome 230 2.0 Array 1390326_at - -
          Rat Genome 230 2.0 Array 1371475_at - -
          Rat Genome 230 2.0 Array 1368943_PM_at - -
          Rat Genome 230 2.0 Array 1368943_at - -
          Rat Genome 230 2.0 Array 1390326_PM_at - -
          Rat Genome 230 2.0 Array 1371475_PM_at - -
          Affymetrix HT Rat Focus Array 1368943_PM_at - -
          Affymetrix HT Rat Focus Array 1368943_at - -
          Affymetrix HT Rat Focus Array 1371475_at - -
          Affymetrix HT Rat Focus Array 1371475_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500002096.rn.1 TC1500000305.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500002097.rn.1 TC1500000305.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500002083.rn.1 TC1500000305.rn.1 RTA TC
          Rat Expression Set 230 1371475_PM_at - -
          Rat Expression Set 230 1368943_at - -
          Rat Expression Set 230 1371475_at - -
          Rat Expression Set 230 1368943_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10779832 - -
          Rat Genome U34 Set AF041066_at - -
          Rat Genome U34 Set rc_AA945613_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21950543 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362410 - -
          RatRef-12 Expression BeadChip ILMN_66191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline