Search
Search
| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024794.1 | NP_001019965.1 | 6 - 7 | 1372 | 125 | |
| XM_008770555.1 | XP_008768777.1 | 6 - 7 | 1745 | 125 | ||
| GenBank mRNA | BC097400.1 | - | 6 - 7 | 1372 | 7453630 | 125 |
Set Membership: |
|