Search
Search
| Gene Symbol: Adck3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013185.1 | NP_001013203.1 | 2 - 3 | 219 | 77 | |
| XM_006250387.1 | XP_006250449.1 | 2 - 3 | 302 | 77 | ||
| GenBank mRNA | BC091388.1 | - | 2 - 3 | 219 | 7321446 | 77 |
Set Membership: |
|