Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01415358_m1
          See other ENAH GE Assays ›
          Gene Symbol:
          Enah
          Gene Name:
          enabled homolog (Drosophila)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 100297421 - 100414361 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01415358_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Enah
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012150.1 NP_001012150.1 11 - 12 1523 125
          XM_006250393.2 XP_006250455.1 12 - 13 2591 125
          XM_006250400.2 XP_006250462.1 13 - 14 1931 125
          XM_006250401.2 XP_006250463.1 12 - 13 1868 125
          XM_008769859.1 XP_008768081.1 14 - 15 2556 125
          XM_008769860.1 XP_008768082.1 12 - 13 3117 125
          XM_008769861.1 XP_008768083.1 12 - 13 2487 125
          GenBank mRNA BC083927.1 - 11 - 12 1523 7135538 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002097.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Enah

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          actin filament organization
          axon guidance
          actin polymerization or depolymerization
          actin cytoskeleton organization
          intracellular transport
          protein binding
          profilin binding
          SH3 domain binding
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P217715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27530 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55557 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69022 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P217715 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P217715 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P217715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P217715 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74471 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103068 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93837 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770418 - -
          Affymetrix Rat Gene 1.0 ST Array 10770417 - -
          Affymetrix Rat Gene 1.0 ST Array 10770424 - -
          Affymetrix Rat Gene 1.0 ST Array 10770413 - -
          Affymetrix Rat Gene 1.0 ST Array 10770421 - -
          Affymetrix Rat Gene 1.0 ST Array 10770423 - -
          Affymetrix Rat Gene 1.0 ST Array 10770428 - -
          Affymetrix Rat Gene 1.0 ST Array 10770426 - -
          Affymetrix Rat Gene 1.0 ST Array 10770416 - -
          Affymetrix Rat Gene 1.0 ST Array 10770427 - -
          Affymetrix Rat Gene 1.0 ST Array 10770420 - -
          Affymetrix Rat Gene 1.0 ST Array 10770419 - -
          Affymetrix Rat Gene 1.0 ST Array 10770414 - -
          Affymetrix Rat Gene 1.0 ST Array 10770415 - -
          Affymetrix Rat Gene 1.1 ST Array 10770412 - -
          Rat Genome 230 2.0 Array 1373535_at - -
          Rat Genome 230 2.0 Array 1375348_PM_at - -
          Rat Genome 230 2.0 Array 1375348_at - -
          Rat Genome 230 2.0 Array 1373535_PM_at - -
          Rat Genome 230 2.0 Array 1396450_PM_at - -
          Rat Genome 230 2.0 Array 1396450_at - -
          Affymetrix HT Rat Focus Array 1375348_at - -
          Affymetrix HT Rat Focus Array 1396450_at - -
          Affymetrix HT Rat Focus Array 1396450_PM_at - -
          Affymetrix HT Rat Focus Array 1375348_PM_at - -
          Affymetrix HT Rat Focus Array 1373535_at - -
          Affymetrix HT Rat Focus Array 1373535_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300014936.rn.1 TC1300002097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014935.rn.1 TC1300002097.rn.1 RTA TC
          Rat Expression Set 230 1373535_at - -
          Rat Expression Set 230 1375348_PM_at - -
          Rat Expression Set 230 1373535_PM_at - -
          Rat Expression Set 230 1375348_at - -
          Rat Expression Set 230 1396450_at - -
          Rat Expression Set 230 1396450_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770412 - -
          Rat Genome U34 Set rc_AA800105_at - -
          Rat Genome U34 Set rc_AA957653_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21728727 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376318 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline