Search
Search
| Gene Symbol: Susd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105982.1 | NP_001099452.1 | 5 - 6 | 893 | 121 | |
| XM_006250375.2 | XP_006250437.1 | 5 - 6 | 932 | 121 | ||
| XM_008769823.1 | XP_008768045.1 | 5 - 6 | 1047 | 121 | ||
| GenBank mRNA | FQ212123.1 | - | 5 - 6 | 877 | 121 | |
Set Membership: |
|