Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416183_m1
          See other PIGA GE Assays ›
          Gene Symbol:
          Piga
          Gene Name:
          phosphatidylinositol glycan anchor biosynthesis, class A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 31786823 - 31799590 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416183_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Piga
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108816.2 NP_001102286.1 4 - 5 494 61
          XM_006256870.2 XP_006256932.1 5 - 6 2744 61
          XM_006256873.2 XP_006256935.1 5 - 6 1322 61
          XM_006256875.2 XP_006256937.1 5 - 6 2864 61
          XM_006256876.1 XP_006256938.1 5 - 6 1378 61
          XM_008773175.1 XP_008771397.1 4 - 5 4042 61
          XM_008773176.1 XP_008771398.1 5 - 6 3725 61
          XM_008773177.1 XP_008771399.1 5 - 6 3313 61
          XM_008773178.1 XP_008771400.1 5 - 6 3781 61
          XM_008773179.1 XP_008771401.1 5 - 6 2789 61
          XM_008773180.1 XP_008771402.1 5 - 6 2845 61
          XM_008773181.1 XP_008771403.1 5 - 6 1846 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001633.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          biological_process
          biosynthetic process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138580 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60367 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138580 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138580 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138585 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138580 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045526 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31196 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937722 - -
          Affymetrix Rat Gene 1.0 ST Array 10937721 - -
          Affymetrix Rat Gene 1.0 ST Array 10937720 - -
          Affymetrix Rat Gene 1.0 ST Array 10937723 - -
          Affymetrix Rat Gene 1.0 ST Array 10937724 - -
          Affymetrix Rat Gene 1.1 ST Array 10937719 - -
          Rat Genome 230 2.0 Array 1396561_x_at - -
          Rat Genome 230 2.0 Array 1396561_PM_x_at - -
          Affymetrix HT Rat Focus Array 1396561_PM_x_at - -
          Affymetrix HT Rat Focus Array 1396561_x_at - -
          Affymetrix Rat Clariom S Assay PSR0X00010555.rn.1 TC0X00001633.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010556.rn.1 TC0X00001633.rn.1 RTA TC
          Rat Expression Set 230 1396561_x_at - -
          Rat Expression Set 230 1396561_PM_x_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937719 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline