Search
Search
| Gene Symbol: Ace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012006.1 | NP_001012006.1 | 4 - 5 | 582 | 90 | |
| GenBank mRNA | AY881244.1 | - | 4 - 5 | 582 | 90 | |
| GQ262788.1 | - | 4 - 5 | 582 | 90 | ||
Set Membership: |
|