Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416312_m1
          See other CTPS2 GE Assays ›
          Gene Symbol:
          Ctps2
          Gene Name:
          CTP synthase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 33383087 - 33522838 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416312_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctps2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034998.2 NP_001030170.1 16 - 17 1635 67
          XM_006256895.2 XP_006256957.1 16 - 17 1637 67
          XM_006256896.2 XP_006256958.1 16 - 17 1925 67
          XM_006256897.2 XP_006256959.1 16 - 17 1758 67
          XM_006256898.2 XP_006256960.1 16 - 17 1719 67
          XM_006256899.2 XP_006256961.1 16 - 17 2090 67
          XM_006256900.2 XP_006256962.1 16 - 17 1733 67
          GenBank mRNA BC085358.1 - 9 - 10 737 67
          BC085949.1 - 16 - 17 1635 7132135 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001649.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctps2

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          glutamine metabolic process
          biological_process
          'de novo' CTP biosynthetic process
          molecular_function
          CTP synthase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P128433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28937 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124408 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36645 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128433 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P128433 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P128433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67955 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20808 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69039 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50352 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85974 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937781 - -
          Affymetrix Rat Gene 1.0 ST Array 10937788 - -
          Affymetrix Rat Gene 1.0 ST Array 10937783 - -
          Affymetrix Rat Gene 1.0 ST Array 10937778 - -
          Affymetrix Rat Gene 1.0 ST Array 10937799 - -
          Affymetrix Rat Gene 1.0 ST Array 10937797 - -
          Affymetrix Rat Gene 1.0 ST Array 10937786 - -
          Affymetrix Rat Gene 1.0 ST Array 10937796 - -
          Affymetrix Rat Gene 1.0 ST Array 10937790 - -
          Affymetrix Rat Gene 1.0 ST Array 10937780 - -
          Affymetrix Rat Gene 1.0 ST Array 10937787 - -
          Affymetrix Rat Gene 1.0 ST Array 10937795 - -
          Affymetrix Rat Gene 1.0 ST Array 10937784 - -
          Affymetrix Rat Gene 1.0 ST Array 10937789 - -
          Affymetrix Rat Gene 1.0 ST Array 10937794 - -
          Affymetrix Rat Gene 1.0 ST Array 10937792 - -
          Affymetrix Rat Gene 1.0 ST Array 10937791 - -
          Affymetrix Rat Gene 1.0 ST Array 10937782 - -
          Affymetrix Rat Gene 1.0 ST Array 10937793 - -
          Affymetrix Rat Gene 1.0 ST Array 10937779 - -
          Affymetrix Rat Gene 1.0 ST Array 10937785 - -
          Affymetrix Rat Gene 1.1 ST Array 10937777 - -
          Rat Genome 230 2.0 Array 1389135_PM_at - -
          Rat Genome 230 2.0 Array 1374482_at - -
          Rat Genome 230 2.0 Array 1374482_PM_at - -
          Rat Genome 230 2.0 Array 1389135_at - -
          Affymetrix HT Rat Focus Array 1374482_PM_at - -
          Affymetrix HT Rat Focus Array 1374482_at - -
          Affymetrix HT Rat Focus Array 1389135_PM_at - -
          Affymetrix HT Rat Focus Array 1389135_at - -
          Affymetrix Rat Clariom S Assay PSR0X00010691.rn.1 TC0X00001649.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010690.rn.1 TC0X00001649.rn.1 RTA TC
          Rat Expression Set 230 1374482_at - -
          Rat Expression Set 230 1374482_PM_at - -
          Rat Expression Set 230 1389135_at - -
          Rat Expression Set 230 1389135_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937777 - -
          Rat Genome U34 Set rc_AA893059_at - -
          Rat Genome U34 Set rc_AI009560_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21824884 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline