Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416539_m1
          See other CLDN19 GE Assays ›
          Gene Symbol:
          Cldn19
          Gene Name:
          claudin 19
          Gene Aliases:
          claudin-19
          Chromosome Location:
          Chr.5: 138298630 - 138307982 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416539_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cldn19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008514.1 NP_001008514.1 3 - 4 558 110
          XM_006238756.2 XP_006238818.1 3 - 4 1030 110
          XM_008763997.1 XP_008762219.1 4 - 5 1015 110
          GenBank mRNA AF497645.1 - 3 - 4 474 110
          BC079172.1 - 3 - 4 558 7129992 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001348.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cldn19

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          neuronal action potential propagation
          apical junction assembly
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138415 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138420 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138415 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138415 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138420 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138420 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138415 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1047578 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10871566 - -
          Affymetrix Rat Gene 1.0 ST Array 10871562 - -
          Affymetrix Rat Gene 1.0 ST Array 10871565 - -
          Affymetrix Rat Gene 1.0 ST Array 10871567 - -
          Affymetrix Rat Gene 1.0 ST Array 10871563 - -
          Affymetrix Rat Gene 1.0 ST Array 10871564 - -
          Affymetrix Rat Gene 1.0 ST Array 10871568 - -
          Affymetrix Rat Gene 1.0 ST Array 10871569 - -
          Affymetrix Rat Gene 1.1 ST Array 10871561 - -
          Rat Genome 230 2.0 Array 1390411_at - -
          Rat Genome 230 2.0 Array 1390411_PM_at - -
          Affymetrix HT Rat Focus Array 1390411_PM_at - -
          Affymetrix HT Rat Focus Array 1390411_at - -
          Affymetrix Rat Clariom S Assay PSR0500009403.rn.1 TC0500001348.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500009406.rn.1 TC0500001348.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500009404.rn.1 TC0500001348.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500009405.rn.1 TC0500001348.rn.1 RTA TC
          Rat Expression Set 230 1390411_at - -
          Rat Expression Set 230 1390411_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871561 - -
          Rat Genome U34 Set rc_AI136542_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21476447 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356908 - -
          RatRef-12 Expression BeadChip ILMN_60483 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline