Search
Search
| Gene Symbol: Hspb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131002.1 | NP_001124474.1 | 5 - 6 | 457 | 83 | |
| XM_006238524.2 | XP_006238586.2 | 5 - 6 | 463 | 83 | ||
| XM_006238525.1 | XP_006238587.1 | 7 - 8 | 722 | 83 | ||
| XM_006238526.2 | XP_006238588.1 | 7 - 8 | 691 | 83 | ||
| XM_006238527.2 | XP_006238589.1 | 6 - 7 | 624 | 83 | ||
| XM_006238530.2 | XP_006238592.1 | 5 - 6 | 441 | 83 | ||
| XM_008763898.1 | XP_008762120.1 | 7 - 8 | 708 | 83 | ||
| XM_008763899.1 | XP_008762121.1 | 4 - 5 | 380 | 83 | ||
| XM_008763900.1 | XP_008762122.1 | 6 - 7 | 607 | 83 | ||
| GenBank mRNA | BC162011.1 | - | 5 - 6 | 457 | 9091347 | 83 |
Set Membership: |
|