Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01416912_m1
          See other HSPB11 GE Assays ›
          Gene Symbol:
          Hspb11
          Gene Name:
          heat shock protein family B (small), member 11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 126856215 - 126882913 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01416912_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hspb11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001131002.1 NP_001124474.1 6 - 7 493 153
          XM_006238524.2 XP_006238586.2 6 - 7 499 153
          XM_006238525.1 XP_006238587.1 8 - 9 758 153
          XM_006238526.2 XP_006238588.1 8 - 9 727 153
          XM_006238527.2 XP_006238589.1 7 - 8 660 153
          XM_006238530.2 XP_006238592.1 6 - 7 477 153
          XM_008763898.1 XP_008762120.1 8 - 9 744 153
          XM_008763899.1 XP_008762121.1 5 - 6 416 153
          XM_008763900.1 XP_008762122.1 7 - 8 643 153
          GenBank mRNA BC162011.1 - 6 - 7 493 9091347 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001141.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          skeletal system development
          smoothened signaling pathway
          heart development
          lung development
          cilium assembly
          left/right axis specification

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P434893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157240 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P434893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P434893 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P434893 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P434893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51225 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870666 - -
          Affymetrix Rat Gene 1.0 ST Array 10870662 - -
          Affymetrix Rat Gene 1.0 ST Array 10870665 - -
          Affymetrix Rat Gene 1.0 ST Array 10870664 - -
          Affymetrix Rat Gene 1.1 ST Array 10870661 - -
          Rat Genome 230 2.0 Array 1397940_PM_at - -
          Rat Genome 230 2.0 Array 1386016_PM_at - -
          Rat Genome 230 2.0 Array 1386016_at - -
          Rat Genome 230 2.0 Array 1397940_at - -
          Rat Genome 230 2.0 Array 1379853_at - -
          Rat Genome 230 2.0 Array 1379853_PM_at - -
          Affymetrix HT Rat Focus Array 1379853_PM_at - -
          Affymetrix HT Rat Focus Array 1379853_at - -
          Affymetrix HT Rat Focus Array 1397940_at - -
          Affymetrix HT Rat Focus Array 1386016_at - -
          Affymetrix HT Rat Focus Array 1386016_PM_at - -
          Affymetrix HT Rat Focus Array 1397940_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500007561.rn.1 TC0500001141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007560.rn.1 TC0500001141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500007563.rn.1 TC0500001141.rn.1 RTA TC
          Rat Expression Set 230 1386016_PM_at - -
          Rat Expression Set 230 1379853_PM_at - -
          Rat Expression Set 230 1386016_at - -
          Rat Expression Set 230 1379853_at - -
          Rat Expression Set 230 1397940_PM_at - -
          Rat Expression Set 230 1397940_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870661 - -
          Rat Genome U34 Set rc_AA799891_at - -
          Rat Genome U34 Set rc_AA799891_g_at - -
          Rat Genome U34 Set rc_AI058985_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21261576 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline