Search
Search
| Gene Symbol: Rcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017132.2 | NP_058828.2 | 4 - 5 | 789 | 99 | |
| GenBank mRNA | BC061962.1 | - | 4 - 5 | 743 | 5598390 | 99 |
| U15734.1 | - | 4 - 5 | 800 | 99 | ||
Set Membership: |
|