Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01418845_g1
          See other LAMA1 GE Assays ›
          Gene Symbol:
          Lama1
          Gene Name:
          laminin, alpha 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 115916907 - 116042123 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01418845_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lama1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108237.2 NP_001101707.2 62 - 63 9593 157
          GenBank mRNA AB425340.1 - 62 - 63 9593 157
          AB425341.1 - 62 - 63 9593 157

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001309.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelial sheet
          cell adhesion
          cell surface receptor signaling pathway
          biological_process
          tissue development
          regulation of cell adhesion
          regulation of cell migration
          neuron projection development
          establishment of epithelial cell apical/basal polarity
          regulation of embryonic development
          retina development in camera-type eye
          epithelial tube branching involved in lung morphogenesis
          branching involved in salivary gland morphogenesis
          retinal blood vessel morphogenesis
          molecular_function
          receptor binding
          extracellular matrix structural constituent
          glycosphingolipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57732 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49576 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156344 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44554 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19193 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19193 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68301 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925864 - -
          Affymetrix Rat Gene 1.0 ST Array 10925917 - -
          Affymetrix Rat Gene 1.0 ST Array 10925900 - -
          Affymetrix Rat Gene 1.0 ST Array 10925889 - -
          Affymetrix Rat Gene 1.0 ST Array 10925867 - -
          Affymetrix Rat Gene 1.0 ST Array 10925891 - -
          Affymetrix Rat Gene 1.0 ST Array 10925870 - -
          Affymetrix Rat Gene 1.0 ST Array 10925901 - -
          Affymetrix Rat Gene 1.0 ST Array 10925922 - -
          Affymetrix Rat Gene 1.0 ST Array 10925912 - -
          Affymetrix Rat Gene 1.0 ST Array 10925906 - -
          Affymetrix Rat Gene 1.0 ST Array 10925910 - -
          Affymetrix Rat Gene 1.0 ST Array 10925919 - -
          Affymetrix Rat Gene 1.0 ST Array 10925921 - -
          Affymetrix Rat Gene 1.0 ST Array 10925897 - -
          Affymetrix Rat Gene 1.0 ST Array 10925899 - -
          Affymetrix Rat Gene 1.0 ST Array 10925896 - -
          Affymetrix Rat Gene 1.0 ST Array 10925879 - -
          Affymetrix Rat Gene 1.0 ST Array 10925920 - -
          Affymetrix Rat Gene 1.0 ST Array 10925898 - -
          Affymetrix Rat Gene 1.0 ST Array 10925872 - -
          Affymetrix Rat Gene 1.0 ST Array 10925877 - -
          Affymetrix Rat Gene 1.0 ST Array 10925883 - -
          Affymetrix Rat Gene 1.0 ST Array 10925907 - -
          Affymetrix Rat Gene 1.0 ST Array 10925861 - -
          Affymetrix Rat Gene 1.0 ST Array 10925886 - -
          Affymetrix Rat Gene 1.0 ST Array 10925865 - -
          Affymetrix Rat Gene 1.0 ST Array 10925868 - -
          Affymetrix Rat Gene 1.0 ST Array 10925882 - -
          Affymetrix Rat Gene 1.0 ST Array 10925902 - -
          Affymetrix Rat Gene 1.0 ST Array 10925918 - -
          Affymetrix Rat Gene 1.0 ST Array 10925871 - -
          Affymetrix Rat Gene 1.0 ST Array 10925881 - -
          Affymetrix Rat Gene 1.0 ST Array 10925914 - -
          Affymetrix Rat Gene 1.0 ST Array 10925911 - -
          Affymetrix Rat Gene 1.0 ST Array 10925875 - -
          Affymetrix Rat Gene 1.0 ST Array 10925904 - -
          Affymetrix Rat Gene 1.0 ST Array 10925913 - -
          Affymetrix Rat Gene 1.0 ST Array 10925869 - -
          Affymetrix Rat Gene 1.0 ST Array 10925909 - -
          Affymetrix Rat Gene 1.0 ST Array 10925862 - -
          Affymetrix Rat Gene 1.0 ST Array 10925888 - -
          Affymetrix Rat Gene 1.0 ST Array 10925892 - -
          Affymetrix Rat Gene 1.0 ST Array 10925893 - -
          Affymetrix Rat Gene 1.0 ST Array 10925916 - -
          Affymetrix Rat Gene 1.0 ST Array 10925884 - -
          Affymetrix Rat Gene 1.0 ST Array 10925880 - -
          Affymetrix Rat Gene 1.0 ST Array 10925885 - -
          Affymetrix Rat Gene 1.0 ST Array 10925890 - -
          Affymetrix Rat Gene 1.0 ST Array 10925874 - -
          Affymetrix Rat Gene 1.0 ST Array 10925894 - -
          Affymetrix Rat Gene 1.0 ST Array 10925915 - -
          Affymetrix Rat Gene 1.0 ST Array 10925895 - -
          Affymetrix Rat Gene 1.0 ST Array 10925908 - -
          Affymetrix Rat Gene 1.0 ST Array 10925876 - -
          Affymetrix Rat Gene 1.0 ST Array 10925905 - -
          Affymetrix Rat Gene 1.0 ST Array 10925863 - -
          Affymetrix Rat Gene 1.0 ST Array 10925903 - -
          Affymetrix Rat Gene 1.0 ST Array 10925887 - -
          Affymetrix Rat Gene 1.0 ST Array 10925860 - -
          Affymetrix Rat Gene 1.0 ST Array 10925878 - -
          Affymetrix Rat Gene 1.0 ST Array 10925873 - -
          Affymetrix Rat Gene 1.1 ST Array 10925859 - -
          Rat Genome 230 2.0 Array 1392926_at - -
          Rat Genome 230 2.0 Array 1392926_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900011094.rn.1 TC0900001309.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011092.rn.1 TC0900001309.rn.1 RTA TC
          Rat Expression Set 230 1392926_PM_at - -
          Rat Expression Set 230 1392926_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925859 - -
          Rat Genome U34 Set rc_AA875154_at - -
          Rat Genome U34 Set rc_AA955427_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21111637 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361319 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline