Search
Search
| Gene Symbol: Dlgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767428.1 | XP_008765650.1 | 4 - 5 | 1863 | 59 | |
| XM_008767429.1 | XP_008765651.1 | 4 - 5 | 1063 | 59 | ||
| XM_008767430.1 | XP_008765652.1 | 4 - 5 | 1094 | 59 | ||
| XM_008767431.1 | XP_008765653.1 | 4 - 5 | 1088 | 59 | ||
| XM_008767432.1 | XP_008765654.1 | 4 - 5 | 1538 | 59 | ||
| XM_008767433.1 | XP_008765655.1 | 4 - 5 | 750 | 59 | ||
| GenBank mRNA | AB003594.1 | - | 4 - 5 | 736 | 59 | |
| FQ212368.1 | - | 4 - 5 | 852 | 59 | ||
| U67137.1 | - | 6 - 7 | 1883 | 59 | ||
Set Membership: |
|