Search Thermo Fisher Scientific
Gene Symbol: Babam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006964.1 | NP_001006965.1 | 4 - 5 | 664 | 72 | |
XM_006252857.2 | XP_006252919.1 | 4 - 5 | 688 | 72 | ||
GenBank mRNA | BC083685.1 | - | 4 - 5 | 664 | 7189038 | 72 |
FQ214886.1 | - | 4 - 5 | 655 | 72 | ||
FQ233705.1 | - | 4 - 5 | 675 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |