Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01420496_gH
          See other KLF2 GE Assays ›
          Gene Symbol:
          Klf2
          Gene Name:
          Kruppel-like factor 2
          Gene Aliases:
          Lklf
          Chromosome Location:
          Chr.16: 19223087 - 19225037 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01420496_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007684.1 NP_001007685.1 2 - 3 887 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001339.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klf2

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          in utero embryonic development
          transcription, DNA-templated
          negative regulation of interleukin-6 production
          erythrocyte homeostasis
          response to laminar fluid shear stress
          multicellular organism growth
          positive regulation of transcription from RNA polymerase II promoter in response to stress
          regulation of gene expression, epigenetic
          erythrocyte maturation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of protein metabolic process
          Type I pneumocyte differentiation
          cellular response to hydrogen peroxide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to organic cyclic compound
          cellular response to cycloheximide
          cellular response to laminar fluid shear stress
          cellular response to peptide
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P472661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123147 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P472661 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P472661 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P472661 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P472661 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790672 - -
          Affymetrix Rat Gene 1.0 ST Array 10790671 - -
          Affymetrix Rat Gene 1.0 ST Array 10790673 - -
          Affymetrix Rat Gene 1.1 ST Array 10790670 - -
          Rat Genome 230 2.0 Array 1376569_at - -
          Rat Genome 230 2.0 Array 1386040_PM_at - -
          Rat Genome 230 2.0 Array 1394068_PM_x_at - -
          Rat Genome 230 2.0 Array 1375248_PM_at - -
          Rat Genome 230 2.0 Array 1394068_x_at - -
          Rat Genome 230 2.0 Array 1386041_a_at - -
          Rat Genome 230 2.0 Array 1386041_PM_a_at - -
          Rat Genome 230 2.0 Array 1386040_at - -
          Rat Genome 230 2.0 Array 1376569_PM_at - -
          Rat Genome 230 2.0 Array 1375248_at - -
          Affymetrix HT Rat Focus Array 1376569_at - -
          Affymetrix HT Rat Focus Array 1394068_x_at - -
          Affymetrix HT Rat Focus Array 1376569_PM_at - -
          Affymetrix HT Rat Focus Array 1394068_PM_x_at - -
          Affymetrix HT Rat Focus Array 1375248_PM_at - -
          Affymetrix HT Rat Focus Array 1386040_at - -
          Affymetrix HT Rat Focus Array 1375248_at - -
          Affymetrix HT Rat Focus Array 1386041_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386040_PM_at - -
          Affymetrix HT Rat Focus Array 1386041_a_at - -
          Affymetrix Rat Clariom S Assay PSR1600010047.rn.1 TC1600001339.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10790670 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376868 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline