Search
Search

St3gal5| Gene Symbol: St3gal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031337.2 | NP_112627.2 | 5 - 6 | 1152 | 135 | |
| XM_006236676.1 | XP_006236738.1 | 4 - 5 | 813 | 135 | ||
| XM_006236677.1 | XP_006236739.1 | 5 - 6 | 955 | 135 | ||
| XM_006236679.2 | XP_006236741.1 | 4 - 5 | 946 | 135 | ||
| XM_006236680.1 | XP_006236742.1 | 4 - 5 | 813 | 135 | ||
| XM_006236682.1 | XP_006236744.1 | 3 - 4 | 640 | 135 | ||
| XM_006236684.2 | XP_006236746.1 | 3 - 4 | 834 | 135 | ||
| XM_008762996.1 | XP_008761218.1 | 4 - 5 | 975 | 135 | ||
| XM_008762997.1 | XP_008761219.1 | 3 - 4 | 863 | 135 | ||
| GenBank mRNA | AB018049.1 | - | 4 - 5 | 818 | 135 | |
| BC078798.1 | - | 5 - 6 | 1152 | 7106217 | 135 | |
Set Membership: |
|