Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01420873_g1
          See other TATDN2 GE Assays ›
          Gene Symbol:
          Tatdn2
          Gene Name:
          TatD DNase domain containing 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 145653300 - 145669343 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01420873_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tatdn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109252.1 NP_001102722.1 7 - 8 2539 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001582.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA catabolic process, endonucleolytic
          DNA catabolic process
          deoxyribonuclease activity
          endodeoxyribonuclease activity, producing 5'-phosphomonoesters

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11902 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117668 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117668 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117668 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P420710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70034 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10857909 - -
          Affymetrix Rat Gene 1.0 ST Array 10857913 - -
          Affymetrix Rat Gene 1.0 ST Array 10857912 - -
          Affymetrix Rat Gene 1.0 ST Array 10857908 - -
          Affymetrix Rat Gene 1.0 ST Array 10857915 - -
          Affymetrix Rat Gene 1.0 ST Array 10857911 - -
          Affymetrix Rat Gene 1.0 ST Array 10857914 - -
          Affymetrix Rat Gene 1.0 ST Array 10857910 - -
          Affymetrix Rat Gene 1.1 ST Array 10857891 - -
          Rat Genome 230 2.0 Array 1374608_PM_at - -
          Rat Genome 230 2.0 Array 1374608_at - -
          Affymetrix HT Rat Focus Array 1374608_at - -
          Affymetrix HT Rat Focus Array 1374608_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011290.rn.1 TC0400001582.rn.1 RTA TC
          Rat Expression Set 230 1374608_PM_at - -
          Rat Expression Set 230 1374608_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857891 - -
          Rat Genome U34 Set rc_AA799581_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21719201 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355729 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline