Search
Search
| Gene Symbol: Camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134468.1 | NP_604463.1 | 9 - 10 | 942 | 78 | |
| XM_006236999.2 | XP_006237061.1 | 10 - 11 | 1611 | 78 | ||
| XM_006237000.2 | XP_006237062.1 | 10 - 11 | 1607 | 78 | ||
| XM_006237001.2 | XP_006237063.1 | 9 - 10 | 1474 | 78 | ||
| XM_006237002.2 | XP_006237064.1 | 9 - 10 | 1401 | 78 | ||
| XM_006237003.2 | XP_006237065.1 | 9 - 10 | 1400 | 78 | ||
| GenBank mRNA | BC071177.1 | - | 9 - 10 | 877 | 7098592 | 78 |
| FQ213485.1 | - | 905 | 78 | |||
| L24907.1 | - | 9 - 10 | 916 | 78 | ||
| L26288.1 | - | 9 - 10 | 942 | 78 | ||
Set Membership: |
|