Search
Search
| Gene Symbol: Crbn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015003.2 | NP_001015003.1 | 7 - 8 | 855 | 88 | |
| GenBank mRNA | AY973953.1 | - | 7 - 8 | 842 | 88 | |
| BC099779.1 | - | 7 - 8 | 840 | 88 | ||
| BC127455.1 | - | 7 - 8 | 847 | 7112865 | 88 | |
Set Membership: |
|