Search
Search
| Gene Symbol: Immt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034928.1 | NP_001030100.1 | 8 - 9 | 973 | 69 | |
| XM_008762981.1 | XP_008761203.1 | 9 - 10 | 1203 | 69 | ||
| XM_008762982.1 | XP_008761204.1 | 9 - 10 | 1203 | 69 | ||
| XM_008762983.1 | XP_008761205.1 | 9 - 10 | 1169 | 69 | ||
| XM_008762984.1 | XP_008761206.1 | 9 - 10 | 1166 | 69 | ||
| XM_008762985.1 | XP_008761207.1 | 9 - 10 | 1169 | 69 | ||
| XM_008762986.1 | XP_008761208.1 | 8 - 9 | 1105 | 69 | ||
| XM_008762987.1 | XP_008761209.1 | 8 - 9 | 1071 | 69 | ||
| XM_008762988.1 | XP_008761210.1 | 8 - 9 | 1068 | 69 | ||
| XM_008762989.1 | XP_008761211.1 | 8 - 9 | 1071 | 69 | ||
| XM_008762990.1 | XP_008761212.1 | 7 - 8 | 978 | 69 | ||
| GenBank mRNA | BC083835.1 | - | 928 | 69 | ||
| BC105841.1 | - | 8 - 9 | 973 | 7454026 | 69 | |
| FQ217458.1 | - | 7 - 8 | 900 | 69 | ||
Set Membership: |
|