Search
Search
| Gene Symbol: Snx22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106832.1 | NP_001100302.1 | 6 - 7 | 522 | 66 | |
| XM_006243304.2 | XP_006243366.1 | 5 - 6 | 537 | 66 | ||
| XM_006243305.2 | XP_006243367.1 | 5 - 6 | 560 | 66 | ||
| XM_008766240.1 | XP_008764462.1 | 6 - 7 | 604 | 66 | ||
Set Membership: |
|