Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Il18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019165.1 | NP_062038.1 | 3 - 4 | 225 | 126 | |
GenBank mRNA | AJ222813.1 | - | 3 - 4 | 225 | 126 | |
AY077842.1 | - | 453 | 126 | |||
AY258448.1 | - | 3 - 4 | 225 | 126 | ||
U77776.1 | - | 3 - 4 | 225 | 126 | ||
U77777.1 | - | 3 - 4 | 225 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |