Search
Search
| Gene Symbol: Fbxo22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037770.1 | NP_001032859.1 | 1 - 2 | 157 | 70 | |
| GenBank mRNA | BC087061.1 | - | 1 - 2 | 187 | 70 | |
| BC107670.1 | - | 1 - 2 | 157 | 7937607 | 70 | |
Set Membership: |
|