Search
Search
| Gene Symbol: Dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199388.1 | NP_955420.1 | 6 - 7 | 1044 | 108 | |
| XM_006242921.2 | XP_006242983.1 | 6 - 7 | 1395 | 108 | ||
| GenBank mRNA | BC063184.1 | - | 6 - 7 | 1044 | 6887995 | 108 |
| FQ209511.1 | - | 4 - 5 | 483 | 108 | ||
| FQ213783.1 | - | 6 - 7 | 1121 | 108 | ||
Set Membership: |
|