Search
Search
| Gene Symbol: Susd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238210.2 | XP_006238272.1 | 12 - 13 | 3480 | 108 | |
| XM_006238211.2 | XP_006238273.1 | 12 - 13 | 3477 | 108 | ||
| XM_006238212.2 | XP_006238274.1 | 12 - 13 | 1868 | 108 | ||
| XM_006238213.2 | XP_006238275.1 | 11 - 12 | 3300 | 108 | ||
Set Membership: |
|