Search
Search
| Gene Symbol: Hsdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025697.1 | NP_001020868.1 | 9 - 10 | 1450 | 79 | |
| XM_008763735.1 | XP_008761957.1 | 10 - 11 | 1385 | 79 | ||
| XM_008763736.1 | XP_008761958.1 | 10 - 11 | 1336 | 79 | ||
| GenBank mRNA | BC097407.1 | - | 9 - 10 | 1450 | 7454038 | 79 |
Set Membership: |
|