Search
Search

Cyp24a1| Gene Symbol: Cyp24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201635.2 | NP_963966.1 | 3 - 4 | 546 | 68 | |
| XM_006235672.2 | XP_006235734.1 | 3 - 4 | 888 | 68 | ||
| GenBank mRNA | BC100059.1 | - | 3 - 4 | 891 | 7129117 | 68 |
| X59506.1 | - | 3 - 4 | 889 | 68 | ||
Set Membership: |
|