Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423218_g1
          See other ADA GE Assays ›
          Gene Symbol:
          Ada
          Gene Name:
          adenosine deaminase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 160115840 - 160139947 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423218_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ada
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130399.2 NP_569083.1 10 - 11 1052 67
          GenBank mRNA AB059655.1 - 10 - 11 976 67
          BC088116.1 - 10 - 11 1052 7384286 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004718.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ada

          Panther Classification:

          Molecular Function -

          deaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          in utero embryonic development
          histamine secretion
          trophectodermal cell differentiation
          liver development
          placenta development
          germinal center B cell differentiation
          positive regulation of germinal center formation
          negative regulation of leukocyte migration
          negative regulation of mature B cell apoptotic process
          adenosine catabolic process
          deoxyadenosine catabolic process
          aging
          purine ribonucleoside monophosphate biosynthetic process
          positive regulation of heart rate
          lung development
          positive regulation of B cell proliferation
          purine nucleotide salvage
          positive regulation of T cell differentiation in thymus
          response to vitamin E
          regulation of cell-cell adhesion mediated by integrin
          T cell activation
          negative regulation of circadian sleep/wake cycle, non-REM sleep
          response to drug
          response to hydrogen peroxide
          negative regulation of apoptotic process
          hypoxanthine salvage
          response to morphine
          regulation of circadian sleep/wake cycle, sleep
          regulation of T cell differentiation
          positive regulation of T cell differentiation
          positive regulation of smooth muscle contraction
          dATP catabolic process
          adenosine metabolic process
          hypoxanthine biosynthetic process
          inosine biosynthetic process
          xanthine biosynthetic process
          positive regulation of alpha-beta T cell differentiation
          lung alveolus development
          Peyer's patch development
          embryonic digestive tract development
          negative regulation of inflammatory response
          positive regulation of calcium-mediated signaling
          positive regulation of T cell receptor signaling pathway
          positive regulation of T cell activation
          negative regulation of adenosine receptor signaling pathway
          negative regulation of penile erection
          negative regulation of thymocyte apoptotic process
          negative regulation of mucus secretion
          purine nucleoside binding
          adenosine deaminase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320260 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61641 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157791 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13102 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13102 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13102 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P434541 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13102 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53554 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5481 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851521 - -
          Affymetrix Rat Gene 1.0 ST Array 10851517 - -
          Affymetrix Rat Gene 1.0 ST Array 10851526 - -
          Affymetrix Rat Gene 1.0 ST Array 10851516 - -
          Affymetrix Rat Gene 1.0 ST Array 10851525 - -
          Affymetrix Rat Gene 1.0 ST Array 10851520 - -
          Affymetrix Rat Gene 1.0 ST Array 10851518 - -
          Affymetrix Rat Gene 1.0 ST Array 10851523 - -
          Affymetrix Rat Gene 1.0 ST Array 10851524 - -
          Affymetrix Rat Gene 1.0 ST Array 10851522 - -
          Affymetrix Rat Gene 1.0 ST Array 10851519 - -
          Affymetrix Rat Gene 1.0 ST Array 10851515 - -
          Affymetrix Rat Gene 1.0 ST Array 10851514 - -
          Affymetrix Rat Gene 1.1 ST Array 10851513 - -
          Rat Genome 230 2.0 Array 1370071_at - -
          Rat Genome 230 2.0 Array 1370071_PM_at - -
          Affymetrix HT Rat Focus Array 1370071_at - -
          Affymetrix HT Rat Focus Array 1370071_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300036131.rn.1 TC0300004718.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300036130.rn.1 TC0300004718.rn.1 RTA TC
          Rat Expression Set 230 1370071_at - -
          Rat Expression Set 230 1370071_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851513 - -
          Rat Genome U34 Set rc_AI102968_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358817 - -
          RatRef-12 Expression BeadChip ILMN_53358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline