Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01423229_m1
          See other HNF4A GE Assays ›
          Gene Symbol:
          Hnf4a
          Gene Name:
          hepatocyte nuclear factor 4, alpha
          Gene Aliases:
          Hnf4, Hnf4alpha
          Chromosome Location:
          Chr.3: 159902441 - 159965003 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01423229_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnf4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270931.1 NP_001257860.1 3 - 4 469 79
          NM_001270933.1 NP_001257862.1 3 - 4 469 79
          NM_022180.2 NP_071516.2 2 - 3 429 79
          XM_006235515.2 XP_006235577.1 2 - 3 424 79
          XM_006235516.2 XP_006235578.1 2 - 3 424 79
          GenBank mRNA CK483260.1 - 2 - 3 258 79
          D10554.1 - 2 - 3 305 79
          EF193390.1 - 2 - 3 250 79
          EF193392.1 - 2 - 3 231 79
          FJ608816.1 - 3 - 4 469 79
          FJ608817.1 - 3 - 4 469 79
          FJ608818.1 - 3 - 4 469 79
          FM042560.1 - 2 - 3 434 79
          X57133.1 - 2 - 3 320 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002259.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnf4a

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          ornithine metabolic process
          lipid metabolic process
          acyl-CoA metabolic process
          xenobiotic metabolic process
          establishment of tissue polarity
          sex differentiation
          blood coagulation
          negative regulation of cell proliferation
          response to glucose
          regulation of gastrulation
          regulation of lipid metabolic process
          signal transduction involved in regulation of gene expression
          cell differentiation
          negative regulation of cell growth
          intracellular receptor signaling pathway
          regulation of microvillus assembly
          negative regulation of protein import into nucleus, translocation
          response to drug
          negative regulation of tyrosine phosphorylation of Stat5 protein
          glucose homeostasis
          steroid hormone mediated signaling pathway
          negative regulation of sequence-specific DNA binding transcription factor activity
          cell-cell junction organization
          positive regulation of gluconeogenesis
          positive regulation of fatty acid biosynthetic process
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          regulation of insulin secretion
          response to cAMP
          lipid homeostasis
          phospholipid homeostasis
          SMAD protein signal transduction
          triglyceride homeostasis
          negative regulation of activation of JAK2 kinase activity
          positive regulation of cholesterol homeostasis
          fatty-acyl-CoA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          receptor binding
          fatty acid binding
          protein binding
          transcription factor binding
          zinc ion binding
          palmitoyl-CoA hydrolase activity
          protein homodimerization activity
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline