Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424412_g1
          See other OBP1F GE Assays ›
          Gene Symbol:
          Obp1f
          Gene Name:
          odorant binding protein I f
          Gene Aliases:
          Obp-1f
          Chromosome Location:
          Chr.X: 45271742 - 45284357 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424412_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Obp1f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138903.1 NP_620258.1 555 86
          GenBank mRNA J03093.1 - 555 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Obp1f

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          sensory perception of smell
          transporter activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P524642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P524642 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P524642 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P524642 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P524642 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208151 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73069 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933776 - -
          Affymetrix Rat Gene 1.0 ST Array 10933775 - -
          Affymetrix Rat Gene 1.0 ST Array 10933779 - -
          Affymetrix Rat Gene 1.0 ST Array 10938123 - -
          Affymetrix Rat Gene 1.0 ST Array 10933773 - -
          Affymetrix Rat Gene 1.0 ST Array 10938125 - -
          Affymetrix Rat Gene 1.0 ST Array 10933774 - -
          Affymetrix Rat Gene 1.0 ST Array 10938124 - -
          Affymetrix Rat Gene 1.0 ST Array 10938120 - -
          Affymetrix Rat Gene 1.0 ST Array 10938122 - -
          Affymetrix Rat Gene 1.1 ST Array 10938119 - -
          Affymetrix Rat Gene 1.1 ST Array 10933772 - -
          Rat Genome 230 2.0 Array 1388014_at - -
          Rat Genome 230 2.0 Array 1388014_PM_at - -
          Affymetrix HT Rat Focus Array 1388014_at - -
          Affymetrix HT Rat Focus Array 1388014_PM_at - -
          Rat Expression Set 230 1388014_PM_at - -
          Rat Expression Set 230 1388014_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933772 - -
          Affymetrix Rat Gene 1.0 ST Array 10938119 - -
          Rat Genome U34 Set J03093_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22327055 - -
          ABI Rat Genome Survey Microarray 22341362 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369449 - -
          RatRef-12 Expression BeadChip ILMN_61849 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline