Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01424757_m1
See other ADCY9 GE Assays ›
Gene Symbol:
Adcy9
Gene Name:
adenylate cyclase 9
Gene Aliases:
-
Chromosome Location:
Chr.10: 11392701 - 11515406 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01424757_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Adcy9
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_008767525.1
XP_008765747.1
10 - 11
3144
85
XM_008767526.1
XP_008765748.1
10 - 11
3087
85
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Rattus norvegicus
1700
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
cAMP biosynthetic process
intracellular signal transduction
nucleotide binding
adenylate cyclase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_43_P22744
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_43_P22744
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_43_P22744
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_43_P22744
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P1009001
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P135944
-
-
Array Name
Probe IDs
Targets
Target Type
Rat Genome 230 2.0 Array
1374419_PM_at
-
-
Rat Genome 230 2.0 Array
1395630_at
-
-
Rat Genome 230 2.0 Array
1381063_at
-
-
Rat Genome 230 2.0 Array
1374419_at
-
-
Rat Genome 230 2.0 Array
1381063_PM_at
-
-
Rat Genome 230 2.0 Array
1395630_PM_at
-
-
Affymetrix HT Rat Focus Array
1381063_at
-
-
Affymetrix HT Rat Focus Array
1374419_at
-
-
Affymetrix HT Rat Focus Array
1374419_PM_at
-
-
Affymetrix HT Rat Focus Array
1381063_PM_at
-
-
Rat Expression Set 230
1395630_PM_at
-
-
Rat Expression Set 230
1395630_at
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
21882170
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1357162
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline