Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tvp23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108263.1 | NP_001101733.1 | 3 - 4 | 671 | 62 | |
XM_006245724.2 | XP_006245786.1 | 3 - 4 | 836 | 62 | ||
XM_006245726.2 | XP_006245788.1 | 4 - 5 | 583 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |