Search
Search
| Gene Symbol: Nubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009619.1 | NP_001009619.1 | 8 - 9 | 740 | 70 | |
| GenBank mRNA | BC088221.1 | - | 8 - 9 | 740 | 7321552 | 70 |
| FQ214074.1 | - | 7 - 8 | 684 | 70 | ||
| FQ220638.1 | - | 8 - 9 | 753 | 70 | ||
| FQ222868.1 | - | 8 - 9 | 757 | 70 | ||
| FQ229330.1 | - | 8 - 9 | 756 | 70 | ||
Set Membership: |
|