Search
Search
| Gene Symbol: Serbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145086.1 | NP_659554.1 | 6 - 7 | 1014 | 154 | |
| XM_006236616.1 | XP_006236678.1 | 6 - 7 | 1107 | 154 | ||
| XM_006236617.1 | XP_006236679.1 | 6 - 7 | 1089 | 154 | ||
| XM_006236618.1 | XP_006236680.1 | 6 - 7 | 1044 | 154 | ||
| GenBank mRNA | AF388527.1 | - | 6 - 7 | 1020 | 154 | |
| BC079337.1 | - | 6 - 7 | 1014 | 7106027 | 154 | |
| FQ213573.1 | - | 6 - 7 | 1103 | 154 | ||
| FQ214321.1 | - | 6 - 7 | 1041 | 154 | ||
| FQ223736.1 | - | 6 - 7 | 1062 | 154 | ||
| FQ225988.1 | - | 6 - 7 | 154 | |||
| FQ227901.1 | - | 2 - 3 | 598 | 154 | ||
| FQ228026.1 | - | 6 - 7 | 1025 | 154 | ||
| FQ230135.1 | - | 5 - 6 | 951 | 154 | ||
| FQ230382.1 | - | 6 - 7 | 755 | 154 | ||
| FQ231329.1 | - | 6 - 7 | 729 | 154 | ||
| FQ231840.1 | - | 6 - 7 | 775 | 154 | ||
| FQ232288.1 | - | 6 - 7 | 1017 | 154 | ||
| FQ232894.1 | - | 6 - 7 | 1036 | 154 | ||
| FQ233179.1 | - | 6 - 7 | 1035 | 154 | ||
| FQ233457.1 | - | 6 - 7 | 1086 | 154 | ||
| FQ233487.1 | - | 6 - 7 | 711 | 154 | ||
| FQ233911.1 | - | 6 - 7 | 1018 | 154 | ||
| FQ234284.1 | - | 6 - 7 | 991 | 154 | ||
| FQ234534.1 | - | 6 - 7 | 1018 | 154 | ||
| FQ235054.1 | - | 6 - 7 | 711 | 154 | ||
Set Membership: |
|