Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01425445_m1
          See other HTRA2 GE Assays ›
          Gene Symbol:
          Htra2
          Gene Name:
          HtrA serine peptidase 2
          Gene Aliases:
          Prss25
          Chromosome Location:
          Chr.4: 113883671 - 113886833 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01425445_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htra2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106599.2 NP_001100069.1 6 - 7 1145 122
          GenBank mRNA BC087588.1 - 5 - 6 1019 122
          BC158760.1 - 6 - 7 1145 7103492 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003163.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          ceramide metabolic process
          mitochondrion organization
          adult walking behavior
          adult locomotory behavior
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to herbicide
          positive regulation of cell death
          pentacyclic triterpenoid metabolic process
          forebrain development
          cellular response to heat
          cellular response to interferon-beta
          regulation of multicellular organism growth
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular protein catabolic process
          neuron development
          negative regulation of cell death
          cellular response to retinoic acid
          cellular response to growth factor stimulus
          intrinsic apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1018464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532553 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33923 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1018464 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1018464 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1018464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1018464 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863481 - -
          Affymetrix Rat Gene 1.0 ST Array 10863484 - -
          Affymetrix Rat Gene 1.0 ST Array 10863479 - -
          Affymetrix Rat Gene 1.0 ST Array 10863482 - -
          Affymetrix Rat Gene 1.0 ST Array 10863483 - -
          Affymetrix Rat Gene 1.0 ST Array 10863480 - -
          Affymetrix Rat Gene 1.0 ST Array 10863478 - -
          Affymetrix Rat Gene 1.1 ST Array 10863477 - -
          Rat Genome 230 2.0 Array 1367478_PM_at - -
          Rat Genome 230 2.0 Array 1367478_at - -
          Affymetrix HT Rat Focus Array 1367478_at - -
          Affymetrix HT Rat Focus Array 1367478_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400022341.rn.1 TC0400003163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400022342.rn.1 TC0400003163.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400022344.rn.1 TC0400003163.rn.1 RTA TC
          Rat Expression Set 230 1367478_at - -
          Rat Expression Set 230 1367478_PM_at - -
          Rat Expression Set 230 1367478_at - -
          Rat Expression Set 230 1367478_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863477 - -
          Rat Genome U34 Set rc_AI179329_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22187890 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline