Search
Search
| Gene Symbol: Slc6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024371.2 | NP_077347.1 | 11 - 12 | 1479 | 98 | |
| XM_006237059.1 | XP_006237121.1 | 12 - 13 | 1688 | 98 | ||
| XM_006237060.2 | XP_006237122.1 | 12 - 13 | 3908 | 98 | ||
| GenBank mRNA | M59742.1 | - | 11 - 12 | 1479 | 98 | |
Set Membership: |
|