Search
Search
| Gene Symbol: Elmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013087.1 | NP_001013105.1 | 4 - 5 | 474 | 69 | |
| XM_006236667.2 | XP_006236729.1 | 4 - 5 | 474 | 69 | ||
| XM_006236669.1 | XP_006236731.1 | 3 - 4 | 426 | 69 | ||
| XM_008762978.1 | XP_008761200.1 | 3 - 4 | 436 | 69 | ||
| GenBank mRNA | BC083623.1 | - | 4 - 5 | 474 | 7134106 | 69 |
Set Membership: |
|