Search
Search
| Gene Symbol: Zfp638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107868.1 | NP_001101338.1 | 10 - 11 | 2563 | 99 | |
| XM_006236749.2 | XP_006236811.1 | 10 - 11 | 2706 | 99 | ||
| XM_006236750.2 | XP_006236812.1 | 10 - 11 | 2706 | 99 | ||
| XM_006236751.2 | XP_006236813.1 | 10 - 11 | 2706 | 99 | ||
| XM_006236753.2 | XP_006236815.1 | 10 - 11 | 2706 | 99 | ||
| XM_006236754.2 | XP_006236816.1 | 10 - 11 | 2725 | 99 | ||
Set Membership: |
|